3-Bromonaphtho[2,3-b]benzofuran 98%

Supplier: Ambeed
Warning

A964168-1G A964168-250MG A964168-5G
77269-088EA 23 USD
77269-088 77269-090 77269-092
3-Bromonaphtho[2,3-b]benzofuran 98%
3-Bromonaphtho[2,3-b]benzofuran

Precaución: for Research Use Only

Formula: C₁₆H₉BrO
MDL Number: MFCD32012085
CAS Number: 1256544-32-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR