3-Aminopropan-1-ol 98%

Supplier: Ambeed
Danger

A963628-25G A963628-100G
77282-068EA 12 USD
77282-068 77618-644
3-Aminopropan-1-ol 98%
3-Aminopropan-1-ol

Precaución: for Research Use Only

Formula: C₃H₉NO
MDL Number: MFCD00008223
CAS Number: 156-87-6
UN: 2735
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR