2,3-Dimethoxyphenol 98%

Supplier: Ambeed
Warning

A963595-100G A963595-1G A963595-25G A963595-5G
77256-868EA 376 USD
77256-868 77256-870 77256-872 77256-874
2,3-Dimethoxyphenol 98%
2,3-Dimethoxyphenol

Precaución: for Research Use Only

Formula: C₈H₁₀O₃
MDL Number: MFCD00008366
CAS Number: 5150-42-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR