2,4,5-Trifluorobenzenesulfonyl chloride 98%

Supplier: Ambeed
Danger

Synonyms: 2,4,5-Trifluorobenzene-1-sulphonyl chloride

A962961-10G A962961-1G A962961-5G
77256-862EA 27.81 USD
77256-862 77256-864 77256-866
2,4,5-Trifluorobenzenesulfonyl chloride 98%
2,4,5-Trifluorobenzenesulphonyl chloride

Precaución: for Research Use Only

Formula: C₆H₂ClF₃O₂S
MW: 230.59 g/mol
MDL Number: MFCD00077600
CAS Number: 220227-21-4
UN: 1760
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR