4-Hydroxy-3-(trifluoromethoxy)benzonitrile 97%

Supplier: Ambeed
Warning

A962345-5G A962345-250MG
77256-860EA 453 USD
77256-860 77747-720
4-Hydroxy-3-(trifluoromethoxy)benzonitrile 97%
4-Hydroxy-3-(trifluoromethoxy)benzonitrile

Precaución: for Research Use Only

Formula: C₈H₄F₃NO₂
MDL Number: MFCD18397473
CAS Number: 1093397-72-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR