1-(2,2-Difluoroethyl)piperazine dihydrochloride 98%

Supplier: Ambeed
Warning

A961647-100MG A961647-1G A961647-250MG
77244-930EA 42 USD
77244-930 77244-932 77244-934
1-(2,2-Difluoroethyl)piperazine dihydrochloride 98%
1-(2,2-Difluoroethyl)piperazine dihydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₂F₂N₂·2ClH
MDL Number: MFCD25371148
CAS Number: 1225276-98-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR