(+)-4,5-Bis[hydroxy(diphenyl)methyl]-2,2-dimethyl-1,3-dioxolane ≥97%, ee 99%

Supplier: Ambeed
Warning

A960844-100G A960844-10G A960844-1G A960844-25G A960844-5G
77294-354EA 290.01 USD
77294-354 77294-356 77294-358 77294-360 77294-362
(+)-4,5-Bis[hydroxy(diphenyl)methyl]-2,2-dimethyl-1,3-dioxolane ≥97%, ee 99%
(+)-4,5-Bis[hydroxy(diphenyl)methyl]-2,2-dimethyl-1,3-dioxolane

Precaución: for Research Use Only

Formula: C₃₁H₃₀O₄
MDL Number: MFCD00010079
CAS Number: 93379-49-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR