2-Heptadecylimidazole 98+%

Supplier: Ambeed
Warning

A960809-100G A960809-25G A960809-500G
77269-082EA 31 USD
77269-082 77269-084 77269-086
2-Heptadecylimidazole 98+%
2-Heptadecylimidazole

Precaución: for Research Use Only

Formula: C₂₀H₃₈N₂
MDL Number: MFCD00038620
CAS Number: 23328-87-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR