Methanesulfonato(tricyclohexylphosphine)(2'-amino-1,1'-biphenyl-2-yl)palladium(II) 98%

Supplier: Ambeed
Warning

A960777-100MG A960777-1G A960777-5G A960777-250MG A960777-25G
77256-850EA 16.48 USD
77256-850 77256-852 77618-642 77747-718 77747-719
Methanesulfonato(tricyclohexylphosphine)(2'-amino-1,1'-biphenyl-2-yl)palladium(II) 98%
Methanesulfonato(tricyclohexylphosphine)(2'-amino-1,1'-biphenyl-2-yl)palladium(II)

Precaución: for Research Use Only

Formula: C₃₁H₄₆NO₃PPdS
MW: 650.16 g/mol
MDL Number: MFCD22417235
CAS Number: 1445086-12-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR