(R)-1-{(R)-2-[Di(2-furyl)phosphino]ferrocenyl}ethylbis(2-methylphenyl)phosphine ≥98%, ee 99%

Supplier: Ambeed
Warning

A959636-1G A959636-250MG
77256-846EA 667.44 USD
77256-846 77256-848
(R)-1-{(R)-2-[Di(2-furyl)phosphino]ferrocenyl}ethylbis(2-methylphenyl)phosphine ≥98%, ee 99%
(R)-1-{(R)-2-[Di(2-furyl)phosphino]ferrocenyl}ethylbis(2-methylphenyl)phosphine

Precaución: for Research Use Only

Formula: C₃₄H₃₂FeO₂P₂
MW: 590.41 g/mol
MDL Number: MFCD08561168
CAS Number: 849924-73-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR