(S)-1-[(S)-2-(Diphenylphosphino)ferrocenyl]ethyldi(3,5-xylyl)phosphine 98%

Supplier: Ambeed
Warning

A959182-100MG A959182-1G A959182-250MG A959182-5G
77256-840EA 21.63 USD
77256-840 77256-842 77256-844 77618-640
(S)-1-[(S)-2-(Diphenylphosphino)ferrocenyl]ethyldi(3,5-xylyl)phosphine 98%
(S)-1-[(S)-2-(Diphenylphosphino)ferrocenyl]ethyldi(3,5-xylyl)phosphine

Precaución: for Research Use Only

Formula: C₄₀H₄₀FeP₂
MW: 638.54 g/mol
MDL Number: MFCD08561148
CAS Number: 223121-07-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR