4-Chlorobenzamidoxime 98%

Supplier: Ambeed
Danger

Synonyms: 4-Chloro-N'-hydroxybenzenecarboximidamide, 4-Chloro-N-hydroxybenzimidamide

A959159-25G A959159-5G
77269-078EA 132.87 USD
77269-078 77269-080
4-Chlorobenzamidoxime 98%
4-Chlorobenzamidoxime

Precaución: for Research Use Only

Formula: C₇H₇ClN₂O
MW: 170.6 g/mol
MDL Number: MFCD00029674
CAS Number: 5033-28-3
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR