3-(4-Chlorophenyl)cyclobutanone 97%

Supplier: Ambeed
Warning

A959054-100MG A959054-250MG
77269-074EA 85.49 USD
77269-074 77269-076
3-(4-Chlorophenyl)cyclobutanone 97%
3-(4-Chlorophenyl)cyclobutanone

Precaución: for Research Use Only

Formula: C₁₀H₉ClO
MW: 180.63 g/mol
MDL Number: MFCD04038980
CAS Number: 152714-07-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR