2,5-Dichloro-N-(2-(isopropylthio)phenyl)pyrimidin-4-amine 97%

Supplier: Ambeed
Warning

A956236-100G A956236-1G A956236-25G A956236-5G
77256-816EA 222.48 USD
77256-816 77256-818 77256-820 77256-822
2,5-Dichloro-N-(2-(isopropylthio)phenyl)pyrimidin-4-amine 97%
2,5-Dichloro-N-(2-(isopropylthio)phenyl)pyrimidin-4-amine

Precaución: for Research Use Only

Formula: C₁₃H₁₃Cl₂N₃S
MW: 314.23 g/mol
MDL Number: MFCD28347652
CAS Number: 1632485-14-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR