5-Oxopiperazine-2-carboxylic acid hydrochloride 95%
Supplier: Ambeed
|
Warning
|
A955401-100MG
A955401-1G
A955401-250MG
77269-068EA
71.07
USD
77269-068
77269-070
77269-072
5-Oxopiperazine-2-carboxylic acid hydrochloride 95%
5-Oxopiperazine-2-carboxylic acid hydrochloride
Precaución: for Research Use Only
Formula:
C₅H₉ClN₂O₃ MW: 180.58 g/mol |
MDL Number:
MFCD28123956 CAS Number: 1922857-21-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...