(2-Amino-4,5-dimethoxyphenyl)(p-tolyl)methanone 97%

Supplier: Ambeed
Warning

A954770-1G A954770-250MG
77244-918EA 60 USD
77244-918 77244-920
(2-Amino-4,5-dimethoxyphenyl)(p-tolyl)methanone 97%
(2-Amino-4,5-dimethoxyphenyl)(p-tolyl)methanone

Precaución: for Research Use Only

Formula: C₁₆H₁₇NO₃
MDL Number: MFCD06739582
CAS Number: 391251-83-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR