2,6-Bis(bromomethyl)pyridine 99%

Supplier: Ambeed
Danger

A954553-25G A954553-5G A954553-1G
77256-812EA 612.85 USD
77256-812 77256-814 77573-888
2,6-Bis(bromomethyl)pyridine 99%
2,6-Bis(bromomethyl)pyridine

Precaución: for Research Use Only

Formula: C₇H₇Br₂N
MW: 264.95 g/mol
MDL Number: MFCD00191795
CAS Number: 7703-74-4
UN: 1759
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR