2-(4-Morpholino)-5-nitropyridine 98%

Supplier: Ambeed
Warning

Synonyms: 4-(5-Nitropyridin-2-yl)morpholine

A953888-1G A953888-25G A953888-5G
77294-346EA 11.33 USD
77294-346 77294-348 77294-350
2-(4-Morpholino)-5-nitropyridine 98%
2-(4-Morpholino)-5-nitropyridine

Precaución: for Research Use Only

Formula: C₉H₁₁N₃O₃
MW: 209.2019 g/mol
MDL Number: MFCD00023308
CAS Number: 26820-62-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR