1,4-Dihydroxy-2,6-dimethoxybenzene 95%

Supplier: Ambeed
Warning

Synonyms: 2,6-Dimethoxybenzene-1,4-diol

A952878-1G A952878-250MG A952878-5G
77256-802EA 35.02 USD
77256-802 77256-804 77256-806
1,4-Dihydroxy-2,6-dimethoxybenzene 95%
1,4-Dihydroxy-2,6-dimethoxybenzene

Precaución: for Research Use Only

Formula: C₈H₁₀O₄
MW: 170.17 g/mol
MDL Number: MFCD00017160
CAS Number: 15233-65-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR