2-Chloro-N,N-dimethylacetamide 97%

Supplier: Ambeed
Danger

A952101-10G A952101-1G A952101-25G A952101-5G
77269-054EA 10.3 USD
77269-054 77269-056 77269-058 77269-060
2-Chloro-N,N-dimethylacetamide 97%
2-Chloro-N,N-dimethylacetamide

Precaución: for Research Use Only

Formula: C₄H₈ClNO
MDL Number: MFCD00039360
CAS Number: 2675-89-0
UN: 3265
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR