E7820 98%

Supplier: Ambeed
Warning

Synonyms: 3-Cyano-N-(3-cyano-4-methyl-1H-indol-7-yl)benzenesulphonamide

A950982-10MG A950982-1MG A950982-5MG
77269-048EA 58.71 USD
77269-048 77269-050 77269-052
E7820 98%
E7820

Precaución: for Research Use Only

Formula: C₁₇H₁₂N₄O₂S
MW: 336.37 g/mol
MDL Number: MFCD09833234
CAS Number: 289483-69-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR