2-Chloro-5-(trifluoromethoxy)phenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (2-Chloro-5-(trifluoromethoxy)phenyl)boronic acid, 2-Chloro-5-(trifluoromethoxy)benzeneboronic acid

A950600-100MG A950600-250MG
77282-036EA 5 USD
77282-036 77282-038
2-Chloro-5-(trifluoromethoxy)phenylboronic acid 98%
2-Chloro-5-(trifluoromethoxy)phenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₅BClF₃O₃
MDL Number: MFCD06656267
CAS Number: 1022922-16-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR