1-Tridecanol 98%

Supplier: Ambeed
Warning

Synonyms: 1-Tridecanol

A950537-5G A950537-100G A950537-10G A950537-25G
77244-906EA 27.81 USD
77244-906 77618-624 77618-626 77618-628
1-Tridecanol 98%
1-Tridecanol

Precaución: for Research Use Only

Formula: H₃C(CH₂)₁₁CH₂OH
MW: 200.36 g/mol
MDL Number: MFCD00004756
CAS Number: 112-70-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR