1,1-Bis((2R,5R)-2,5-diethylphospholano)ferrocene(cyclooctadiene)rhodium(I) 98%
Supplier: Ambeed
|
Warning
|
A950013-100MG
A950013-1G
A950013-250MG
77256-796EA
132
USD
77256-796
77256-798
77256-800
1,1-Bis((2R,5R)-2,5-diethylphospholano)ferrocene(cyclooctadiene)rhodium(I) 98%
1,1-Bis((2R,5R)-2,5-diethylphospholano)ferrocene(cyclooctadiene)rhodium(I)
Precaución: for Research Use Only
Formula:
C₃₄H₄₂FeP₂Rh |
MDL Number:
MFCD17018832 CAS Number: 162412-90-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...