2,4,6-Trifluorobenzyl alcohol 98%

Supplier: Ambeed
Warning

A949797-1G A949797-250MG A949797-25G A949797-5G
77256-788EA 11 USD
77256-788 77256-790 77256-792 77256-794
2,4,6-Trifluorobenzyl alcohol 98%
2,4,6-Trifluorobenzyl alcohol

Precaución: for Research Use Only

Formula: C₇H₅F₃O
MDL Number: MFCD00061207
CAS Number: 118289-07-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR