(S)-1-{(S)-2-[Di(1-naphthyl)phosphino]ferrocenyl}ethyldi(3,5-xylyl)phosphine ≥98%, ee 99%

Supplier: Ambeed
Warning

A949783-100MG A949783-250MG
77256-784EA 25 USD
77256-784 77256-786
(S)-1-{(S)-2-[Di(1-naphthyl)phosphino]ferrocenyl}ethyldi(3,5-xylyl)phosphine ≥98%, ee 99%
(S)-1-{(S)-2-[Di(1-naphthyl)phosphino]ferrocenyl}ethyldi(3,5-xylyl)phosphine

Precaución: for Research Use Only

Formula: C₄₈H₄₄FeP₂
MDL Number: MFCD08561163
CAS Number: 851308-41-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR