Methyl-2-(2-fluorophenyl)acetate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-fluorophenylacetate, Methyl 2-(2-fluorophenyl)acetate

A949741-100G A949741-10G A949741-25G A949741-5G
77282-028EA 71 USD
77282-028 77282-030 77282-032 77282-034
Methyl-2-(2-fluorophenyl)acetate 98%
Methyl-2-(2-fluorophenyl)acetate

Precaución: for Research Use Only

Formula: C₉H₉FO₂
MDL Number: MFCD00800605
CAS Number: 57486-67-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR