2-Hydrazino-4,5-dihydro-1H-imidazole hydrobromide 95%

Supplier: Ambeed
Warning

A949404-10G A949404-1G A949404-25G A949404-5G
77269-040EA 65 USD
77269-040 77269-042 77269-044 77269-046
2-Hydrazino-4,5-dihydro-1H-imidazole hydrobromide 95%
2-Hydrazino-4,5-dihydro-1H-imidazole hydrobromide

Precaución: for Research Use Only

Formula: C₃H₉BrN₄
MDL Number: MFCD00012694
CAS Number: 55959-84-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR