1-Bromooctane 99%

Supplier: Ambeed
Danger

Synonyms: 1-Octyl bromide

A949395-25G A949395-100G
77244-904EA 5.15 USD
77244-904 77552-758
1-Bromooctane 99%
1-Bromooctane

Precaución: for Research Use Only

Formula: BrH₂C(CH₂)₆CH₃
MW: 193.13 g/mol
MDL Number: MFCD00000276
CAS Number: 111-83-1
UN: 3082
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR