2-(4-Methylphenyl)ethanol 98%
Supplier: Ambeed
|
Warning
|
A949252-100G
A949252-10G
A949252-1G
A949252-25G
A949252-5G
77244-894EA
111
USD
77244-894
77244-896
77244-898
77244-900
77244-902
2-(4-Methylphenyl)ethanol 98%
2-(4-Methylphenyl)ethanol
Precaución: for Research Use Only
Formula:
C₉H₁₂O |
MDL Number:
MFCD00002904 CAS Number: 699-02-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...