2-Formyl-5-methoxyphenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 5-Methoxy-2-formylphenylboronic acid

A948960-1G A948960-250MG A948960-5G
77256-778EA 50.47 USD
77256-778 77256-780 77256-782
2-Formyl-5-methoxyphenylboronic acid 98%
2-Formyl-5-methoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₉BO₄
MW: 179.9657 g/mol
MDL Number: MFCD03001335
CAS Number: 40138-18-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR