4-Methyl-2-(4-pyridinyl)-5-thiazolecarboxylic acid 98%

Supplier: Ambeed
Warning

Synonyms: 4-Methyl-2-(pyridin-4-yl)thiazole-5-carboxylic acid

A948581-1G A948581-250MG A948581-5G
77294-338EA 78.28 USD
77294-338 77294-340 77294-342
4-Methyl-2-(4-pyridinyl)-5-thiazolecarboxylic acid 98%
4-Methyl-2-(4-pyridinyl)-5-thiazolecarboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₈N₂O₂S
MW: 220.2477 g/mol
MDL Number: MFCD00171757
CAS Number: 144060-98-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR