Chloro(1,5-hexadiene)rhodium(I) dimer 98%
Supplier: Ambeed
|
Danger
|
A948518-100MG
A948518-1G
77256-774EA
119
USD
77256-774
77256-776
Chloro(1,5-hexadiene)rhodium(I) dimer 98%
Chloro(1,5-hexadiene)rhodium(I) dimer
Precaución: for Research Use Only
Formula:
C₁₂H₂₀Cl₂Rh₂ |
MDL Number:
MFCD00064595 CAS Number: 32965-49-4 UN: 1759 ADR: 8,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...