2-Amino-[1,1'-biphenyl]-4,4'-dicarboxylic acid 97%

Supplier: Ambeed
Warning

A947968-100MG A947968-1G A947968-250MG A947968-5G
77269-032EA 10 USD
77269-032 77269-034 77269-036 77269-038
2-Amino-[1,1'-biphenyl]-4,4'-dicarboxylic acid 97%
2-Amino-[1,1'-biphenyl]-4,4'-dicarboxylic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₁NO₄
MDL Number: MFCD32644766
CAS Number: 1240557-01-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR