4-Fluorobenzenesulphonyl chloride 98%

Supplier: Ambeed
Danger

Synonyms: 4-Fluorobenzene-1-sulphonyl chloride

A947771-100G A947771-10G A947771-25G A947771-500G A947771-5G
77294-328EA 49 USD
77294-328 77294-330 77294-332 77294-334 77294-336
4-Fluorobenzenesulphonyl chloride 98%
4-Fluorobenzenesulphonyl chloride

Precaución: for Research Use Only

Formula: C₆H₄ClFO₂S
MDL Number: MFCD00007438
CAS Number: 349-88-2
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR