4'-Fluoroacetanilide 98%

Supplier: Ambeed
Warning

A946810-100G A946810-25G A946810-500G
77294-322EA 35 USD
77294-322 77294-324 77294-326
4'-Fluoroacetanilide 98%
4'-Fluoroacetanilide

Precaución: for Research Use Only

Formula: C₈H₈FNO
MDL Number: MFCD00016335
CAS Number: 351-83-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR