2,6-Difluorobenzenesulfonamide 96%

Supplier: Ambeed
Danger

A946713-25G A946713-5G
77256-762EA 60 USD
77256-762 77256-764
2,6-Difluorobenzenesulfonamide 96%
2,6-Difluorobenzenesulphonamide

Precaución: for Research Use Only

Formula: C₆H₅F₂NO₂S
MDL Number: MFCD00729101
CAS Number: 60230-37-7
UN: 1759
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR