1,1,1-Tris(hydroxymethyl)propane 95%

Supplier: Ambeed
Warning

Synonyms: 1,1,1-Trimethylolpropane, Propylidynetrimethanol, Trimethylol propane, Propylidynetris(methanol), 2-Ethyl-2-(hydroxymethyl)-1,3-propanediol

A946602-1000G A946602-100G A946602-500G
77269-026EA 27.81 USD
77269-026 77269-028 77269-030
1,1,1-Tris(hydroxymethyl)propane 95%
1,1,1-Tris(hydroxymethyl)propane

Precaución: for Research Use Only

Formula: C₆H₁₄O₃
MW: 134.18 g/mol
MDL Number: MFCD00004694
CAS Number: 77-99-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR