3-Bromopropionaldehyde dimethylacetal 97%

Supplier: Ambeed
Danger

Synonyms: 3-Bromo-1,1-dimethoxypropane

A946329-1G A946329-250MG A946329-10G A946329-25G A946329-5G
77282-018EA 9.27 USD
77282-018 77282-020 77552-752 77552-754 77749-774
3-Bromopropionaldehyde dimethylacetal 97%
3-Bromopropionaldehyde dimethylacetal

Precaución: for Research Use Only

Formula: BrH₂CCH₂CH(OCH₃)₂
MW: 183.05 g/mol
MDL Number: MFCD00013245
CAS Number: 36255-44-4
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR