3-Amino-9-ethyl carbazole (AEC) 95%

Supplier: Ambeed
Warning

Synonyms: 9-Ethylcarbazol-3-ylamine, 9-Ethyl-9H-carbazol-3-amine, AEC, 50X, AEC, 3-Amino-9-ethylcarbazole

A945894-1G A945894-250MG A945894-5G
77256-756EA 10.3 USD
77256-756 77256-758 77256-760
3-Amino-9-ethyl carbazole (AEC) 95%
3-Amino-9-ethyl carbazole (AEC)

Precaución: for Research Use Only

Formula: C₁₄H₁₄N₂
MW: 210.28 g/mol
MDL Number: MFCD00004964
CAS Number: 132-32-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR