trans-2-(4-Chlorophenyl)vinylboronic acid 96%

Supplier: Ambeed
Warning

Synonyms: (E)-(4-Chlorostyryl)boronic acid

A945299-100MG A945299-1G A945299-250MG A945299-5G
77244-886EA 14 USD
77244-886 77244-888 77244-890 77244-892
trans-2-(4-Chlorophenyl)vinylboronic acid 96%
trans-2-(4-Chlorophenyl)vinylboronic acid

Precaución: for Research Use Only

Formula: C₈H₈BClO₂
MW: 182.42 g/mol
MDL Number: MFCD02093767
CAS Number: 154230-29-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR