2-Bromodiphenylamine 98%

Supplier: Ambeed
Warning

A944744-1G A944744-250MG A944744-25G A944744-5G
77269-018EA 11 USD
77269-018 77269-020 77269-022 77269-024
2-Bromodiphenylamine 98%
2-Bromodiphenylamine

Precaución: for Research Use Only

Formula: C₁₂H₁₀BrN
MDL Number: MFCD01851817
CAS Number: 61613-22-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR