Terephthalimidamide dihydrochloride 97%

Supplier: Ambeed
Warning

A944650-100MG A944650-1G A944650-250MG
77244-872EA 16 USD
77244-872 77244-874 77244-876
Terephthalimidamide dihydrochloride 97%
Terephthalimidamide dihydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₂Cl₂N₄
MDL Number: MFCD00136761
CAS Number: 14401-56-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR