3-(2,6-Dichlorophenyl)propionic acid 98%

Supplier: Ambeed
Warning

Synonyms: 3-(2,6-Dichlorophenyl)propanoic acid

A944310-1G A944310-25G A944310-5G
77282-012EA 20.6 USD
77282-012 77282-014 77282-016
3-(2,6-Dichlorophenyl)propionic acid 98%
3-(2,6-Dichlorophenyl)propionic acid

Precaución: for Research Use Only

Formula: C₉H₈Cl₂O₂
MW: 219.07 g/mol
MDL Number: MFCD01310807
CAS Number: 51656-68-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR