2-Mesitylenesulfonyl hydrazide 95%

Supplier: Ambeed
Danger

Synonyms: 2,4,6-Trimethylbenzenesulphonohydrazide, 2,4,6-Trimethylbenzenesulphonyl Hydrazide

A943439-100G A943439-1G A943439-25G A943439-5G
77256-748EA 375.95 USD
77256-748 77256-750 77256-752 77256-754
2-Mesitylenesulfonyl hydrazide 95%
2-Mesitylenesulfonyl hydrazide

Precaución: for Research Use Only

Formula: C₉H₁₄N₂O₂S
MW: 214.28 g/mol
MDL Number: MFCD00007585
CAS Number: 16182-15-3
UN: 1325
ADR: 4.1,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR