2,4,6-Trimethylaniline 98%

Supplier: Ambeed
Danger

A943272-1G A943272-25G A943272-100G
77256-746EA 5 USD
77256-746 77552-750 77569-664
2,4,6-Trimethylaniline 98%
2,4,6-Trimethylaniline

Precaución: For research use only.

Formula: C₉H₁₃N
MDL Number: MFCD00007740
CAS Number: 88-05-1
UN: 2810
ADR: 6.1,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR