2-Amino-2'-deoxyadenosine 95%

Supplier: Ambeed
Warning

A941875-1G A941875-250MG A941875-25G A941875-5G
77256-738EA 17 USD
77256-738 77256-740 77256-742 77256-744
2-Amino-2'-deoxyadenosine 95%
2-Amino-2'-deoxyadenosine

Precaución: for Research Use Only

Formula: C₁₀H₁₄N₆O₃
MDL Number: MFCD00047240
CAS Number: 4546-70-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR