(1S)-1-[Bis(4-methoxy-3,5-dimethylphenyl)phosphino]-2-[(1S)-1-(dicyclohexylphosphino)ethyl]ferrocene ≥98%, ee 99%

Supplier: Ambeed
Warning

A941189-100MG A941189-250MG A941189-50MG
77256-732EA 103 USD
77256-732 77256-734 77256-736
(1S)-1-[Bis(4-methoxy-3,5-dimethylphenyl)phosphino]-2-[(1S)-1-(dicyclohexylphosphino)ethyl]ferrocene ≥98%, ee 99%
(1S)-1-[Bis(4-methoxy-3,5-dimethylphenyl)phosphino]-2-[(1S)-1-(dicyclohexylphosphino)ethyl]ferrocene

Precaución: for Research Use Only

Formula: C₄₂H₅₆FeO₂P₂
MW: 710.7 g/mol
MDL Number: MFCD08561152
CAS Number: 849923-88-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR