4-(3-((5-(4-Nitrophenyl)furan-2-yl)methylene)-2-oxo-5-phenyl-2,3-dihydro-1H-pyrrol-1-yl)benzoic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
4E1RCat
A940826-10MG
A940826-1MG
A940826-50MG
A940826-5MG
77244-852EA
44
USD
77244-852
77244-854
77244-856
77244-858
4-(3-((5-(4-Nitrophenyl)furan-2-yl)methylene)-2-oxo-5-phenyl-2,3-dihydro-1H-pyrrol-1-yl)benzoic acid 98%
4-(3-((5-(4-Nitrophenyl)furan-2-yl)methylene)-2-oxo-5-phenyl-2,3-dihydro-1H-pyrrol-1-yl)benzoic acid
Precaución: for Research Use Only
Formula:
C₂₈H₁₈N₂O₆ |
MDL Number:
MFCD01931282 CAS Number: 328998-25-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...