4,4'-Diaminooctafluorobiphenyl 98%

Supplier: Ambeed
Warning

Synonyms: 2,2',3,3',5,5',6,6'-Octafluoro-[1,1'-biphenyl]-4,4'-diamine, Octafluorobenzidine

A940400-1G A940400-25G A940400-5G
77294-316EA 29.87 USD
77294-316 77294-318 77294-320
4,4'-Diaminooctafluorobiphenyl 98%
4,4'-Diaminooctafluorobiphenyl

Precaución: for Research Use Only

Formula: C₁₂H₄F₈N₂
MW: 328.17 g/mol
MDL Number: MFCD00007646
CAS Number: 1038-66-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR